Drain stoppers for SAP and JBN baths
Catalog # GRANUWB-DS
Supplier: GRANT INSTRUMENTS
Specifications
- Per:JBN12, JBN18, JBN26, SAP12, SAP18, SAP26, SAP34
- Descrizione:Tappi di scarico per JBN12, JBN18, JBN26, SAP12, SAP18, SAP26, SAP34
- Conf:5
Specifications
About this item
Drain stoppers, recommended when using heat transfer beads, to prevent beads entering the drain hole.