Orders LinkContactUsLinkComponent
Burette stand
Burette stand
Burette stand, with cast iron base (127×203 mm) and zinc plated rod (8×508 mm), 127×203 mm
Codice catalogo HACH56300
Nome del fornitore:   Hach
Burette stand
Codice catalogo HACH56300
Nome del fornitore:  Hach
Burette stand
Burette stand
Burette stand, with cast iron base (127×203 mm) and zinc plated rod (8×508 mm), 127×203 mm
Price
69.90 SFr.
Log in per disponibilità
1 * 1 ST
Product Details & Documents
Heavy black enamel, cast iron base and zinc-plated rod.
Specifiche
  • Descrizione:
    Burette stand, with cast iron base (127×203 mm) and zinc plated rod (8×508 mm)
  • L×L:
    127×203 mm