ContactUsLinkComponent
Anti-DISC1 (FRAG2) Rabbit Polyclonal Antibody
Anti-DISC1 (FRAG2) Rabbit Polyclonal Antibody
  ABFRLF-PA0226
Anti-DISC1 (FRAG2) Rabbit Polyclonal Antibody
  ABFRLF-PA0226
  • Tipo anticorpo:
    Primary
  • Simbolo dell'antigene:
    DISC1
  • Clonalità:
    Policlonale
  • Ospite:
    Rabbit
  • Isotipo:
    IgG
  • Reattività:
    Human
  • Western blot:
    Yes
  • Sinomini antigene:
    FRAG2
  • Temperatura di conservazione:
    Store for 1 year at –20 °C from date of shipment
  • Concentrazione:
    2 mg/ml
  • Immunogeno:
    Recombinant human protein purified from E.coli (His-tagged)
  • Purificazione:
    Affinity purification
  • Taglia:
    100 µl
  • Conf:
    100 µl
Type: Primary
Antigen: DISC1 (FRAG2)
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human