Orders LinkContactUsLinkComponent
Bostwick consistometers
Bostwick consistometers
Supplier:   ENDECOTTS
Bostwick consistometers
Supplier:  ENDECOTTS
Certificates
Bostwick consistometers
Bostwick consistometers
2 Options Available Below
Product Details & Documents
Bostwick consistometers are durable stainless steel instrument for accurately checking laboratory or production samples against consistency, viscosity or flow rate standards.
  • Consistent platform for test
  • Requires up to 100 ml of sample
  • Leveling screws and spirit level enable accurate set up
Used in chemical, paint, cosmetic and food processing industries.
Certifications: R-81294D and ASTM F1080-93
Documents
Product Family Options
Product Information
  • Descrizione
    Availability
    Price
  • Standard consistometer, 240 mm, graduations 48×0,5, 880×355×104 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 2’500.00
    SFr. 2’500.00
    1 * 1 ST
       SFr. 2’500.00
  • Extended consistometer, 300 mm, graduations 60×0,5, 880×414×104 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - SFr. 2’700.00
    SFr. 2’700.00
    1 * 1 ST
       SFr. 2’700.00