Petri dishes, disposable, square, Nunc™ Lab-Tek™
Catalog # NUNC4021
Supplier: Thermo Fisher Scientific
Specifications
- Sterilità:Sterili
- Versione:Square, 100×15 mm, vented, without stacking ring
- Environmentally Preferable:Produzione a basso impatto
- Certificazione ambientale:NAR Certificate of Retirement
- Conf:500
Specifications
About this item
Square Petri dishes for culturing fungi, bacteria and other microorganisms.
Packaging: 20 plates per sealed plastic sleeve.