Extractor for the perforator method, DIN EN 120
NEUB1050-11-1000
- Tipo:NS 71 flange
- Volume matraccio:1000 ml
- Conf:1
Complete assembly includes NS 71 flange, frit, stopcock, cooler, piston, erlenmeyer flask and ball pipe.
Complete assembly includes NS 71 flange, frit, stopcock, cooler, piston, erlenmeyer flask and ball pipe.