Cryo storage box Cryo-Safe™ Cold Box, SP Bel-Art
Codice catalogo 479-1150
Nome del fornitore: Bel-Art Products, a Part of SP
Specifiche
- Per:12 microcentrifuge tubes 1,5 ml
- Dimensioni:117×117×71 mm
- Conf:1
Specifiche
Informazioni su questo articolo
Polystyrene box with polystyrene cover.
- Practical cold storage of perishable samples
- Capacity: twelve 1,5 ml microcentrifuge tubes (tubes not included)
- Holds −15 ºC temperature for 1,5 hours, and maintains 0 ºC for 4 hours when removed from a freezer
The Cryo-Safe™ Cold Box safeguards enzymes and thermolabile substances from thermal degradation. This compact unit is made up of a molded polystyrene box with cover. The interior of the unit is filled with a cryogenic compound and then permanently sealed, to ensure the low temperatures needed to safeguard materials in the cold box.