Order Entry
Switzerland
ContactUsLinkComponent
Anti-RTF1 Rabbit Polyclonal Antibody
Anti-RTF1 Rabbit Polyclonal Antibody
  ABGEAP16561C
 :  Abgent
Anti-RTF1 Rabbit Polyclonal Antibody
  ABGEAP16561C
 :  Abgent

 

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    RTF1 homolog, Paf1/RNA polymerase II complex component
  • Simbolo dell'antigene:
    RTF1
  • Clonalità:
    Policlonale
  • Coniugazione:
    Non coniugato
  • ELISA:
    Yes
  • Ospite:
    Rabbit
  • ImmunoChimica:
    Yes
  • Reattività:
    Human
  • Western blot:
    Yes
  • Epitopo:
    internal
  • ID gene:
    23168
  • Sinomini antigene:
    KIAA0252|GTL7
  • Numero amminoacido:
    482 to 510
  • Tampone di conservazione:
    pH 7,4
  • Peso molecolare:
    80.313 kDa
  • Sequenza:
    KEKERFRKAPPNYAC
  • Temperatura di conservazione:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentrazione:
    0.45
  • Temperatura di spedizione:
    4 °C
  • Immunogeno:
    This RTF1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 482-510 amino acids from the Central region of human RTF1.
  • Purificazione:
    Protein A & Peptide affinity purification
  • Conf:
    400 µl

 

 

Type: Primary
Antigen: RTF1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human