Order Entry
Switzerland
ContactUsLinkComponent
Anti-EEPD1 Rabbit Polyclonal Antibody
Anti-EEPD1 Rabbit Polyclonal Antibody
  ABGEAP18169B
 :  Abgent
Anti-EEPD1 Rabbit Polyclonal Antibody
  ABGEAP18169B
 :  Abgent

 

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    endonuclease/exonuclease/phosphatase family domain containing 1
  • Simbolo dell'antigene:
    EEPD1
  • Clonalità:
    Policlonale
  • Coniugazione:
    Non coniugato
  • ELISA:
    Yes
  • Ospite:
    Rabbit
  • Reattività:
    Human
  • Western blot:
    Yes
  • Epitopo:
    C-terminal
  • ID gene:
    80820
  • Sinomini antigene:
    HSPC107
  • Numero amminoacido:
    438 to 467
  • Tampone di conservazione:
    pH 7,4
  • Peso molecolare:
    62.403 kDa
  • Sequenza:
    PDSNDYDILRKEKFHC
  • Temperatura di conservazione:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentrazione:
    0.5
  • Temperatura di spedizione:
    4 °C
  • Immunogeno:
    This EEPD1 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 438-467 amino acids from the C-terminal region of human EEPD1.
  • Purificazione:
    Protein A & Peptide affinity purification
  • Conf:
    400 µl

 

 

Type: Primary
Antigen: EEPD1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype:
Reactivity: Human