Anti-MTHFSD Rabbit Polyclonal Antibody
ABGEAP17699A
: Abgent
- Tipo anticorpo:Primary
- Tipo di anticorpi:Methenyltetrahydrofolate synthetase domain containing
- Simbolo dell'antigene:MTHFSD
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- ELISA:Yes
- Citometria a flusso:Yes
- Ospite:Rabbit
- Reattività:Human
- Western blot:Yes
- Epitopo:N-terminal
- ID gene:64779
- Numero amminoacido:43 to 69
- Tampone di conservazione:pH 7,4
- Peso molecolare:42.173 kDa
- Sequenza:RTQEVKVDPDKPC
- Temperatura di conservazione:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentrazione:0.45
- Temperatura di spedizione:4 °C
- Immunogeno:This MTHFSD antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 43-69 amino acids from the N-terminal region of human MTHFSD.
- Purificazione:Protein A & Peptide affinity purification
- Conf:400 µl
Type: Primary
Antigen: MTHFSD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human