Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-ITGA7 Rabbit Polyclonal Antibody
Catalog # ABGEAP18839A
Supplier:  Abgent
Anti-ITGA7 Rabbit Polyclonal Antibody
Catalog # ABGEAP18839A
Supplier:  Abgent

Specifications

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    Integrin Alpha 7
  • Simbolo dell'antigene:
    ITGA7
  • Clonalità:
    Policlonale
  • Coniugazione:
    Non coniugato
  • ELISA:
    Yes
  • Ospite:
    Rabbit
  • Reattività:
    Human
  • Western blot:
    Yes
  • Epitopo:
    N-terminal
  • ID gene:
    3679
  • Numero amminoacido:
    229 to 257
  • Tampone di conservazione:
    pH 7,4
  • Peso molecolare:
    128.948 kDa
  • Sequenza:
    GPYEAGGEKEQDPRC
  • Temperatura di conservazione:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentrazione:
    0.3
  • Temperatura di spedizione:
    4 °C
  • Immunogeno:
    This ITGA7 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 229-257 amino acids from the N-terminal region of human ITGA7.
  • Purificazione:
    Protein A & Peptide affinity purification
  • Conf:
    400 µl

Specifications

About this item

Type: Primary
Antigen: ITGA7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human