Order Entry
Switzerland
ContactUsLinkComponent
Anti-PIGP Rabbit Polyclonal Antibody
Anti-PIGP Rabbit Polyclonal Antibody
Catalog # ABGEAP16809A
Supplier:  Abgent
Anti-PIGP Rabbit Polyclonal Antibody
Catalog # ABGEAP16809A
Supplier:  Abgent

Specifications

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    phosphatidylinositol glycan anchor biosynthesis, class P
  • Simbolo dell'antigene:
    PIGP
  • Clonalità:
    Policlonale
  • Coniugazione:
    Non coniugato
  • ELISA:
    Yes
  • Ospite:
    Rabbit
  • Reattività:
    Human
  • Western blot:
    Yes
  • Epitopo:
    N-terminal
  • ID gene:
    51227
  • Sinomini antigene:
    DSRC|DSCR5|DCRC-S|DCRC
  • Numero amminoacido:
    2 to 30
  • Tampone di conservazione:
    pH 7,4
  • Peso molecolare:
    18.059 kDa
  • Sequenza:
    RLSKAPGKMVENSPC
  • Temperatura di conservazione:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentrazione:
    0.7
  • Temperatura di spedizione:
    4 °C
  • Immunogeno:
    This PIGP antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 2-30 amino acids from the N-terminal region of human PIGP.
  • Purificazione:
    Protein A & Peptide affinity purification
  • Conf:
    400 µl

Specifications

About this item

Type: Primary
Antigen: PIGP
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human