Order Entry
Switzerland
ContactUsLinkComponent
Anti-TRGC2 Rabbit Polyclonal Antibody
Anti-TRGC2 Rabbit Polyclonal Antibody
Catalog # ABGEAP19092A
Supplier:  Abgent
Anti-TRGC2 Rabbit Polyclonal Antibody
Catalog # ABGEAP19092A
Supplier:  Abgent

Specifications

  • Tipo anticorpo:
    Primary
  • Simbolo dell'antigene:
    TRGC2
  • Clonalità:
    Policlonale
  • Coniugazione:
    Non coniugato
  • ELISA:
    Yes
  • Ospite:
    Rabbit
  • ImmunoChimica:
    Yes
  • Reattività:
    Human
  • Western blot:
    Yes
  • Epitopo:
    N-terminal
  • Numero amminoacido:
    1 to 30
  • Tampone di conservazione:
    pH 7,4
  • Peso molecolare:
    21.554 kDa
  • Sequenza:
    DKQLDADVSPKPC
  • Temperatura di conservazione:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentrazione:
    0.5
  • Temperatura di spedizione:
    4 °C
  • Immunogeno:
    This TRGC2 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 1-30 amino acids from the N-terminal region of human TRGC2.
  • Purificazione:
    Protein A & Peptide affinity purification
  • Conf:
    400 µl

Specifications

About this item

Type: Primary
Antigen: TRGC2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-terminal
Host: Rabbit
Isotype:
Reactivity: Human