Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-His-tagged IKK-gamma Mouse Monoclonal Antibody [clone: 72C627]
  CAYM13931-1
Anti-His-tagged IKK-gamma Mouse Monoclonal Antibody [clone: 72C627]
  CAYM13931-1

 

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    His-tagged IKK-gamma
  • Simbolo dell'antigene:
    NF-kappaB
  • Clonalità:
    Monoclonale
  • Clone:
    72C627
  • Coniugazione:
    Non coniugato
  • Ospite:
    Mouse
  • Isotipo:
    IgG
  • Reattività:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitopo:
    Full length
  • Assorbimento incrociato:
    No
  • Forma:
    Liquid
  • Tampone di conservazione:
    PBS, containing 0,05% BSA and 0,05% sodium azide
  • Temperatura di conservazione:
    –20 °C
  • Temperatura di spedizione:
    –20 °C
  • Immunogeno:
    His-tagged full-length human IKK-gamma
  • Purificazione:
    Purified by Protein G chromatography
  • Taglia:
    1 Ea
  • Conf:
    1

 

 

Nuclear Factor-κB (NF-κB) is sequestered in the cytoplasm by the IκB family of inhibitory proteins that mask the nuclear localization signal of NF-κB, thereby preventing translocation of NF-κB to the nucleus. External stimuli such as tumor necrosis factor or other cytokines result in phosphorylation and degradation of IκB, releasing NF-κB dimers. NF-κB dimers subsequently translocate to the nucleus and activate target genes. Synthesis of IκBα is autoregulated. IκB proteins are phosphorylated by IκB kinase complex consisting of at least three proteins, IKK1/α, IKK2/β, and IKK3/γ. IKKγ preferentially interacts with IKKβ and is required for activation of IKK complex. IKKγ is also known as NF-κB essential modulator (NEMO). The human T-cell leukemia virus type I Tax oncoprotein that activates NF-κB binds neither to IKKα nor IKKβ, but complexes directly with IKKγ. This suggests that IKKγ may be a key molecule acting as an adapter for oncoprotein specific signaling to IKKα and IKKβ.

  • Starting concentration for WB is 2 µg/ml
  • For immunochemical analysis of IKKγ

Type: Primary
Antigen: His-tagged IKK-gamma
Clonality: Monoclonal
Clone: 72C627
Conjugation: unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse