Anti-E2F7 Rabbit Polyclonal Antibody
100 μl
Codice catalogo ORIGTA321958
Nome del fornitore: OriGene
Vai a
Product Details & Documents
Recommended Dilutions: ELISA: 1:1000-5000; Immunochemistry: 1:50-200
Type: Primary
Antigen: E2F7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Documents
TA321958
Scheda dati
Specifiche
- Taglia:100 μl
- Reattività:Human|Rat|Mouse
- Assorbimento incrociato:No
- Immunogeno:Synthetic peptide corresponding to a region derived from 53-66 amino acids of human E2F transcription factor 7
- Purificazione:purified by antigen affinity chromatography
- Tampone di conservazione:PBS pH 7,3, 0,05% NaN3, 50% glycerol
- Coniugazione:Non coniugato
- Isotipo:IgG
- Cat. No.:ORIGTA321958
- Forma:Liquid
- Numero amminoacido:53 to 66
- Simbolo dell'antigene:E2F7
- Clonalità:Policlonale
- Ospite:Rabbit
- Temperatura di spedizione:–20 °C
- Tipo anticorpo:Primary
- Temperatura di conservazione:–20 °C
- ImmunoChimica:Yes
- ELISA:Yes
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



