Order Entry
Switzerland
ContactUsLinkComponent
Anti-TNFRSF10B Mouse Polyclonal Antibody [clone: DJR2-4 (a.k.a. 7-8)]
Anti-TNFRSF10B Mouse Polyclonal Antibody [clone: DJR2-4 (a.k.a. 7-8)]
Catalog # ORIGTA320492
Supplier:  OriGene
Anti-TNFRSF10B Mouse Polyclonal Antibody [clone: DJR2-4 (a.k.a. 7-8)]
Catalog # ORIGTA320492
Supplier:  OriGene

Specifications

  • Tipo anticorpo:
    Primary
  • Simbolo dell'antigene:
    TNFRSF10B
  • Clonalità:
    Policlonale
  • Clone:
    DJR2-4 (a.k.a. 7-8)
  • Coniugazione:
    Non coniugato
  • Ospite:
    Mouse
  • Reattività:
    Human
  • Assorbimento incrociato:
    No
  • Forma:
    Liquid
  • Tampone di conservazione:
    Aqueous buffer, 0,09% sodium azide, may contain carrier protein/stabilizer
  • Temperatura di conservazione:
    –20 °C
  • Temperatura di spedizione:
    –20 °C
  • Purificazione:
    purified by affinity chromatography
  • Taglia:
    25 μg
  • Conf:
    25 µG

Specifications

About this item

Type: Primary
Antigen: TNFRSF10B
Clonality: Polyclonal
Clone: DJR2-4 (a.k.a. 7-8)
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human