Specifications
- Tipo anticorpo:Primary
- Tipo di anticorpi:Acetylcholinesterase
- Simbolo dell'antigene:ACHE
- Clonalità:Policlonale
- Ospite:Rabbit
- Isotipo:IgG
- Reattività:Human,Rat,Mouse
- Western blot:Yes
- Forma:Lyophilized
- ID gene:43
- Sinomini antigene:N-ACHE|YT|ARACHE|ACEE
- UniProtKB:P22303
- Temperatura di conservazione:At -20˚C for one year. After reconstitution, at 4˚C for one month. It can also be aliquotted and stored frozen at -20˚C for a longer time. Avoid repeated freezing and thawing.
- Immunogeno:A synthetic peptide corresponding to a sequence at the C-terminus of human ACHE (592-614aa SSYMVHWKNQFDHYSKQDRCSDL), different from the related mouse and rat sequences by one amino acid.
- Purificazione:Immunogen affinity purified.
- Taglia:100 µg/vial
- Conf:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Acetylcholinesterase(ACHE) detection. Tested with WB in Human;Mouse;Rat.
ACHE is also known as Acetylcholinesterase. And Acetylcholinesterase hydrolyzes the neurotransmitter, acetylcholine at neuromuscular junctions and brain cholinergic synapses, and thus terminates signal transmission. It is also found on the red blood cell membranes, where it constitutes the Yt blood group antigen. Acetylcholinesterase exists in multiple molecular forms which possess similar catalytic properties, but differ in their oligomeric assembly and mode of cell attachment to the cell surface. It is encoded by the single ACHE gene, and the structural diversity in the gene products arises from alternative mRNA splicing, and post-translational associations of catalytic and structural subunits. The major form of acetylcholinesterase found in brain, muscle and other tissues is the hydrophilic species, which forms disulfide-linked oligomers with collagenous, or lipid-containing structural subunits. The other, alternatively spliced form, expressed primarily in the erythroid tissues, differs at the C-terminal end, and contains a cleavable hydrophobic peptide with a GPI-anchor site. It associates with the membranes through the phosphoinositide (PI) moieties added post-translationally.
Add 0.2ml of distilled water will yield a concentration of 500ug/ml.
Each vial contains 5mg BSA, 0.9mg NaCl, 0.2mg Na2HPO4, 0.05mg NaN3.
WB: The detection limit for ACHE is approximately 0.1ng/lane under reducing conditions.
Tested Species: In-house tested species with positive results.
Other applications have not been tested.
Optimal dilutions should be determined by end users.
Type: Primary
Antigen: ADIPOR1
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Rat