Mortar with pestle
Catalog # 39792.KT
Anbieter: Thermo Fisher Scientific
Technische Daten
- Ø:95 mm
- Conf:1
Technische Daten
Über diesen Artikel
Agate. High quality with flawless grinding surface.
Agate. High quality with flawless grinding surface.