Syringe rack
Syringe rack, holds up to 5 syringes, 700/1700/7000 series syringes
Codice catalogo HAMI204880
Nome del fornitore: HAMILTON BONADUZ
Vai a
Product Details & Documents
This brushed stainless steel rack is used for clean and safe storage of syringes with plastic holder. A lip on the plastic holder protects the syringe against accidental touching and falling down. The distance of the plastic holder prevents the syringe from getting in contact with the metal plate.
- A trough underneath prevents the soiling of worplace with liquid
- Usable as free standing or wall model
Specifiche
- Per:700/1700/7000 series syringes
- Descrizione:Syringe rack, holds up to 5 syringes