ContactUsLinkComponent
Dosing funnels, with spiral conveyor
Dosing funnels, with spiral conveyor
Catalog # WITG1.897.825
Supplier:  witeg Labortechnik
undefined
Dosing funnels, with spiral conveyor
Catalog # WITG1.897.825
Supplier:  witeg Labortechnik
Specifications
  • Dimensione cono:
    29/32 NS
  • Cono femmina:
    29/32 NS
  • Capacità:
    250 ml
  • Conf:
    1
Specifications
About this item
Borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For powdery substances. Horizontal version with spiral conveyor.