Dispenser bucket, Wiper Bowl® Safe & Clean
Catalog # 115-2744
Supplier: Zellstoff-Vertriebs-GmbH & Co. KG
Specifications
- Descrizione:Dispenser bucket, Wiper Bowl® Safe & Clean, empty
- Conf:1
Specifications
About this item
This dispenser bucket made of plastic can be filled with wipe rolls and disinfectant or cleaning solutions. It is ideal for thorough cleaning applications in sensitive areas, e.g. in workshops. If a work area is disinfected simply close the lid and the remaining cloths will remain moist until their next use - for 30 days.
- Refillabl with off-the-shelf disinfectants or cleaning agents
- Economic and hygienic
- Easy dispensing thanks to tear-off perforation