Orders LinkContactUsLinkComponent
RapID™ System
RapID™ System
RapID™ System
RapID™ System
RapID™ System
RapID™ Systems are a complete line of cost-effective manual identification panels for a broad spectrum of microorganisms. RapID™ panels offer easy, one-step, simultaneous inoculation of dehydrated reactants in tray wells to identify anaerobes, coryneform Gram-positive bacilli, enterics, nonfermenters, streptococci, and yeast all within 4 hours. Systems are ideal for manual testing or as confirmation for automated test systems.
  • Rapid, accurate identification of over 400 medically important organisms
  • Simple, one-step inoculation
  • Four-hour aerobic incubation
  • Easy-to-read colour reactions
Documents
Product Family Options
  • Descrizione
    Conf
  • RapID™ One panel
    20
    1 * 20 pezzi
    1 * 20 pezzi - SFr. 185.00
    SFr. 185.00
    1 * 20 pezzi
       SFr. 185.00
  • RapID™ Inoculation Fluid 1 ml
    20
    1 * 20 ST
    1 * 20 ST - SFr. 31.10
    SFr. 31.10
    1 * 20 ST
       SFr. 31.10
  • RapID™ Inoculation Fluid 2 ml
    20
    1 * 20 ST
    1 * 20 ST - SFr. 34.00
    SFr. 34.00
    1 * 20 ST
       SFr. 34.00
  • RapID™ CB Plus panel
    20
    1 * 20 ST
    1 * 20 ST - SFr. 337.00
    SFr. 337.00
    1 * 20 ST
       SFr. 337.00
  • RapID™ Spot Indole reagent
    15 mL
    1 * 15 mL
    1 * 15 mL - SFr. 34.10
    SFr. 34.10
    1 * 15 mL
       SFr. 34.10
  • RapID™ Nitrate A reagent
    15 mL
    1 * 15 mL
    1 * 15 mL - SFr. 30.10
    SFr. 30.10
    1 * 15 mL
       SFr. 30.10
  • RapID™ Nitrate B reagent
    15 mL
    1 * 15 mL
    1 * 15 mL - SFr. 28.90
    SFr. 28.90
    1 * 15 mL
       SFr. 28.90
  • RapID™ NH panel
    20
    1 * 20 ST
    1 * 20 ST - SFr. 220.00
    SFr. 220.00
    1 * 20 ST
       SFr. 220.00
  • RapID™ ANAII panel
    20
    1 * 20 ST
    1 * 20 ST - SFr. 383.00
    SFr. 383.00
    1 * 20 ST
       SFr. 383.00
  • RapID™ Str panel
    20
    1 * 20 ST
    1 * 20 ST - SFr. 364.00
    SFr. 364.00
    1 * 20 ST
       SFr. 364.00
  • RapID™ Ss/U panel
    20
    1 * 20 ST
    1 * 20 ST - SFr. 171.00
    SFr. 171.00
    1 * 20 ST
       SFr. 171.00
  • RapID™ Yeast Plus panel
    20
    1 * 20 ST
    1 * 20 ST - SFr. 332.00
    SFr. 332.00
    1 * 20 ST
       SFr. 332.00
  • RapID™ Eric
    1
    1 * 1 ST
    1 * 1 ST - SFr. 30.20
    SFr. 30.20
    1 * 1 ST
       SFr. 30.20
  • RapID™ Staph Plus System
    20
    1 * 20 ST
    1 * 20 ST - SFr. 442.00
    SFr. 442.00
    1 * 20 ST
       SFr. 442.00