Orders LinkContactUsLinkComponent
Vials
Vials
Vials
Certificates
Vials
35 Options Available Below
Product Details & Documents
Product Family Options
Screw top
Product Information
  • Descrizione
    Capacità
    Colore
    Tappo incluso
    Availability
    Price
  • Vials
    2,00 ml
    ambra
    No
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST
  • Vials
    2,00 ml
    ambra
    No
    Log in for availability
    Log in for availability
    1 * 50.000 ST
    1 * 50.000 ST
  • Vials, cap bundle, LP
    2,00 ml
    ambra
    Yes
    Log in for availability
    Log in for availability
    1 * 50.000 ST
    1 * 50.000 ST
  • Vials, cap bundle, LP
    2,00 ml
    ambra
    Yes
    Log in for availability
    Log in for availability
    1 * 5.000 ST
    1 * 5.000 ST
  • Vials, with write on label
    2,00 ml
    ambra
    No
    Log in for availability
    Log in for availability
    1 * 5.000 ST
    1 * 5.000 ST
  • Vials, SN, with write on label
    2,00 ml
    ambra
    No
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
  • Vials, with write on label
    2,00 ml
    ambra
    No
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST
  • Vials, with write on label
    2,00 ml
    ambra
    No
    Log in for availability
    Log in for availability
    1 * 50.000 ST
    1 * 50.000 ST
  • Vials
    2,00 ml
    trasparente
    No
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST
  • Vials
    2,00 ml
    trasparente
    No
    Log in for availability
    Log in for availability
    1 * 50.000 pezzi
    1 * 50.000 pezzi
  • Vials, cap bundle, LP
    2,00 ml
    trasparente
    Yes
    Log in for availability
    Log in for availability
    1 * 50.000 ST
    1 * 50.000 ST
  • Vials, cap bundle, LP
    2,00 ml
    trasparente
    Yes
    Log in for availability
    Log in for availability
    1 * 5.000 ST
    1 * 5.000 ST
  • Vials, cap bundle, LP, with write on label
    2,00 ml
    trasparente
    Yes
    Log in for availability
    Log in for availability
    1 * 50.000 ST
    1 * 50.000 ST
  • Vials, cap bundle, LP, with write on label
    2,00 ml
    trasparente
    Yes
    Log in for availability
    Log in for availability
    1 * 5.000 ST
    1 * 5.000 ST
  • Vials
    2,00 ml
    trasparente
    No
    Log in for availability
    Log in for availability
    1 * 5.000 ST
    1 * 5.000 ST
  • Vials, with write on label
    2,00 ml
    trasparente
    No
    Log in for availability
    Log in for availability
    1 * 5.000 ST
    1 * 5.000 ST
  • Vials, SN
    2,00 ml
    trasparente
    No
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
  • Vials, with write on label
    2,00 ml
    trasparente
    No
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
Crimp top
Product Information
  • Descrizione
    Capacità
    Colore
    Tappo incluso
    Availability
    Price
  • Vials, cap bundle, LP
    2,00 ml
    ambra
    Yes
    Log in for availability
    Log in for availability
    1 * 50.000 ST
    1 * 50.000 ST
  • Vials, cap bundle, LP
    2,00 ml
    ambra
    Yes
    Log in for availability
    Log in for availability
    1 * 5.000 ST
    1 * 5.000 ST
  • Vials, with write on label
    2,00 ml
    ambra
    No
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST
  • Vials, with write on label
    2,00 ml
    ambra
    No
    Log in for availability
    Log in for availability
    1 * 50.000 ST
    1 * 50.000 ST
  • Vials, LP, with write on label
    2,00 ml
    ambra
    No
    Log in for availability
    Log in for availability
    1 * 5.000 ST
    1 * 5.000 ST
  • Vials, cap bundle, LP
    2,00 ml
    trasparente
    Yes
    Log in for availability
    Log in for availability
    1 * 50.000 ST
    1 * 50.000 ST
  • Vials, cap bundle, LP
    2,00 ml
    trasparente
    Yes
    Log in for availability
    Log in for availability
    1 * 5.000 ST
    1 * 5.000 ST
  • Vials, cap bundle, LP, with write on label
    2,00 ml
    trasparente
    Yes
    Log in for availability
    Log in for availability
    1 * 50.000 ST
    1 * 50.000 ST
  • Vials, cap bundle, LP, with write on label
    2,00 ml
    trasparente
    Yes
    Log in for availability
    Log in for availability
    1 * 5.000 ST
    1 * 5.000 ST
  • Vials, LP
    2,00 ml
    trasparente
    No
    Log in for availability
    Log in for availability
    1 * 5.000 ST
    1 * 5.000 ST
  • Vials, LP, with write on label
    2,00 ml
    trasparente
    No
    Log in for availability
    Log in for availability
    1 * 5.000 ST
    1 * 5.000 ST
  • Vials, SN, with write on label
    2,00 ml
    trasparente
    No
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
  • Vials, wide
    2,00 ml
    trasparente
    No
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST
  • Vials, wide
    2,00 ml
    trasparente
    No
    Log in for availability
    Log in for availability
    1 * 50.000 ST
    1 * 50.000 ST
  • Vials, wide, with write on label
    2,00 ml
    No
    Log in for availability
    Log in for availability
    1 * 10.000 ST
    1 * 10.000 ST
  • Vials, wide, with write on label
    2,00 ml
    No
    Log in for availability
    Log in for availability
    1 * 50.000 ST
    1 * 50.000 ST
This product has been blocked by your organization. Please contact your purchasing department for more information
Snap top
Product Information
  • Descrizione
    Capacità
    Colore
    Tappo incluso
    Availability
    Price
  • Vials, with write on label
    2,00 ml
    trasparente
    No
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST
This product has been blocked by your organization. Please contact your purchasing department for more information