Dewar vessel calorimeter
Catalog # DIDAPTD039501
Supplier: DIDALAB
Specifications
- Descrizione:Dewar vessel calorimeter
- Conf:1
Specifications
About this item
This calorimeter includes an aluminium container of 500 ml, concentric to one large neck Dewar vase. The metal internal vase is supported by plastic ring, laying on the Dewar vessel large neck. The set is protected by a metal casing, equipped with two locking hooks.
White internal plastic cover including a square section opening for the stirring rod, blocked in rotation because of the square section of the rod.
Central closing with Ø 40 mm protection cap, for isolation of foreign bodies after having been introduced into the calorimeter and a thermometer of Ø 8 mm opening.
Cover with rubber washer hold by two metal clamp collars diametrically opposed (outer Ø of 170 mm, height of 215 mm).
Delivery: Supplied with glass beaker 500 ml and handbook.