Gas purifier, 73-099 AA
Product Details & Documents
This wall mounted gas purifier system is designed to purify compressed air and acetylene gas.
It consists of two independent filtration systems, one for compressed air and one for acetylene. The unit also has a flashback arrestor on the acetylene line and a pressure regulator on the compressed air line.
The two-stage air filtration assembly consists of a Balston grade DX coalescing filter and a Balston grade BX coalescing filter. These filters remove oil, water and particulate contamination (99,99% at 0,01 micron) from the compressed air supply.
Specifiche
- Descrizione:Gas purifier, 73-099 AA
- Cat. No.:PERCN9301398
- Gas purificati:Compressed air and acetylene gases
Accessories
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Viton® O-ring for acetylene filter
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Viton® O-ring kit for air filter
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Replacement acetylene filter cartridge
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Replacement air filter cartridge, 1st stageReplacement air filter cartridge, 1st stagePERCN9301710Log in per disponibilità1 * 1 ST - SFr. 85.80
Informazioni sul prodotto
- Codice catalogoDisponibilitàPrice
- Replacement air filter cartridge, 2nd stageReplacement air filter cartridge, 2nd stagePERCN9301711Log in per disponibilità1 * 1 ST - SFr. 85.80