Flow meters, soap bubble
Soap bubble flow meter (two stage) 1 and 10 ml
Codice catalogo PERC00230522
Nome del fornitore: PerkinElmer
Vai a
Product Details & Documents
Two stage soap bubble flow meter.
Informazioni per l'ordine: Supplied with liquid soap, rubber squeeze bulb, plastic tubes and instruction manual.
Specifiche
- Descrizione:Soap bubble flow meter (two stage) 1 and 10 ml
- Seq:02