Volumetric flasks, class A, glass, clear, NS plastic stopper
Catalog # WITG3.664.0030
Supplier: witeg Labortechnik
Specifications
- Cono (NS):10/19
- Capacità:30 ml
- Environmentally Preferable:
- Conf:1
- Tolleranza:0,04 ± ml
Specifications
About this item
DURAN®, borosilicate glass, clear, with PE stopper.
- With acid and alkali resistant ring marks and inscriptions
- DIFFICO stain markings
Certifications: DIN 12664