Reusable earplugs, E-A-R™ Ultrafit™ X
111-1629
: 3M
- Colore:giallo/blu
- Descrizione:Earplugs, with cord, 35 dB, with storage box
- Seq:02
- Taglia:U*
- Conf:50 PAIR
Corded earplugs made from a patented polymer formulation, the comfortable triple flanged tip of these earplugs provides an excellent attenuation of 35 dB SNR, one of the highest amongst pre-moulded earplugs. The earplugs are already pre-formed to ensure optimum fit.
- Triple-flange feature of the earplugs ensures a perfect fit: No size adjustment required
- Fit most ear canals comfortably and provide a high level of attenuation
- Moisture resistant and compatible with other PPE
- Washable and reusable
SNR: 35 dB
: EN 352-2