Order Entry
Switzerland
ContactUsLinkComponent
Anti-USP3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Anti-USP3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # USBIU4099-70B-AP
Supplier:  US Biological
undefined
Anti-USP3 Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase))
Catalog # USBIU4099-70B-AP
Supplier:  US Biological

Specifications

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    Ubiquitin specific peptidase 3
  • Simbolo dell'antigene:
    USP3
  • Clonalità:
    Policlonale
  • Coniugazione:
    AP (Alcalin fosfatasi)
  • ELISA:
    Yes
  • Ospite:
    Rabbit
  • Isotipo:
    IgG
  • Reattività:
    Human
  • Western blot:
    Yes
  • Epitopo:
    N-terminal
  • Forma:
    liquid
  • Tampone di conservazione:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Temperatura di conservazione:
    Cool
  • Temperatura di spedizione:
    Ice
  • Purificazione:
    purified by Protein G chromatography
  • Taglia:
    200 µl
  • Conf:
    200 µl

Specifications

About this item

Recommended dilution: ELISA: 1:1000; Immunochemistry: 1:50-1:100
Do not freeze!

Type: Primary
Antigen: USP3 (ubiquitin specific peptidase 3)
Clonality: polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human