Order Entry
Switzerland
ContactUsLinkComponent
Anti-TES Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Anti-TES Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Catalog # USBI042742-APC
Supplier:  US Biological
Anti-TES Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Catalog # USBI042742-APC
Supplier:  US Biological

Specifications

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    testis derived transcript (3 LIM domains)
  • Simbolo dell'antigene:
    TES
  • Clonalità:
    Policlonale
  • Coniugazione:
    APC (Allofilcocianina)
  • ELISA:
    Yes
  • Ospite:
    Rabbit
  • Isotipo:
    IgG
  • Reattività:
    Human
  • Western blot:
    Yes
  • Epitopo:
    N-terminal
  • Forma:
    liquid
  • Numero amminoacido:
    41 to 70
  • Tampone di conservazione:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Temperatura di conservazione:
    Cool
  • Temperatura di spedizione:
    Ice
  • Applicazioni testate:
    Fluoresence ELISA
  • Purificazione:
    purified by Protein A chromatography
  • Taglia:
    200 µl
  • Conf:
    200 µl

Specifications

About this item

Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: TES (testis derived transcript (3 LIM domains))
Clonality: Polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: N-terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human