Order Entry
Switzerland
ContactUsLinkComponent
Anti-Interferon alpha Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 278CT1]
Anti-Interferon alpha Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 278CT1]
  USBI031057-FITC
undefined
Anti-Interferon alpha Mouse Monoclonal Antibody (FITC (Fluorescein Isothiocyanate)) [clone: 278CT1]
  USBI031057-FITC

 

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    Interferon alpha
  • Simbolo dell'antigene:
    IFNA
  • Clonalità:
    Monoclonale
  • Clone:
    278CT1
  • Coniugazione:
    FITC (Fluoresceina Isotiocionato)
  • ELISA:
    Yes
  • Ospite:
    Mouse
  • Isotipo:
    IgG1 kappa
  • Reattività:
    Human
  • Western blot:
    Yes
  • Forma:
    liquid
  • Tampone di conservazione:
    PBS, pH 7,2, contains ~0,09% sodium azide
  • Temperatura di conservazione:
    Cool
  • Temperatura di spedizione:
    Ice
  • Purificazione:
    purified by Protein G chromatography
  • Taglia:
    100 µl
  • Conf:
    200 µl

 

 

Recommended dilution: FLISA: 1:1000; Western Blot: 1:100

Type: Primary
Antigen: Interferon alpha
Clonality: Monoclonal
Clone: 278CT1
Conjugation: FITC (Fluorescein)
Epitope:
Host: Mouse
Isotype: IgG1 Kappa
Reactivity: Human