Order Entry
Switzerland
ContactUsLinkComponent
Anti-IgG F(c) Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Anti-IgG F(c) Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
  ROCK709-1017
Anti-IgG F(c) Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
  ROCK709-1017

 

  • Tipo anticorpo:
    Secondary
  • Simbolo dell'antigene:
    IgG F(c)
  • Clonalità:
    Policlonale
  • Coniugazione:
    TRITC (Tetrametillrodamina Isotiocianato)
  • ELISA:
    Yes
  • Citometria a flusso:
    Yes
  • Spostamento del gel:
    Yes
  • Ospite:
    Goat
  • ImmunoChimica:
    Yes
  • ImmunoFluorescenza:
    Yes
  • ImmunoPrecipitazione:
    Yes
  • Reattività:
    Human
  • Western blot:
    Yes
  • Assorbimento incrociato:
    Bovine, Horse, Mouse, Rat
  • ChIP:
    Yes
  • Neutralisation:
    Yes
  • Formato:
    Antibody fraction
  • Forma:
    Lyophilized
  • Frammento anticorpo:
    F(ab')2
  • Tampone di conservazione:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Temperatura di conservazione:
    Cool
  • Concentrazione:
    1,0 mg/ml by UV absorbance at 280 nm
  • Temperatura di spedizione:
    Ice
  • Immunogeno:
    Human IgG F(c) fragment
  • Purificazione:
    purified by immunoaffinity chromatography
  • Taglia:
    1 mg
  • Conf:
    1 mg

 

 

Secondary Goat Anti-IgG F(c) Reacts with Human

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: TRITC (Tetramethylrhodamine Isothiocyanate)
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Human