Order Entry
Switzerland
ContactUsLinkComponent
Anti-IgA/IgG/IgM Goat Polyclonal Antibody (Peroxidase)
Anti-IgA/IgG/IgM Goat Polyclonal Antibody (Peroxidase)
Catalog # ROCK618-103-130
Anti-IgA/IgG/IgM Goat Polyclonal Antibody (Peroxidase)
Catalog # ROCK618-103-130

Specifications

  • Tipo anticorpo:
    Secondary
  • Simbolo dell'antigene:
    IgA/IgG/IgM
  • Clonalità:
    Policlonale
  • Coniugazione:
    Perossidasi
  • ELISA:
    Yes
  • Citometria a flusso:
    Yes
  • Spostamento del gel:
    Yes
  • Ospite:
    Goat
  • ImmunoChimica:
    Yes
  • ImmunoPrecipitazione:
    Yes
  • Reattività:
    Ferret
  • Western blot:
    Yes
  • Assorbimento incrociato:
    none
  • ChIP:
    Yes
  • Neutralisation:
    Yes
  • Formato:
    Antibody fraction
  • Forma:
    Lyophilized
  • Tampone di conservazione:
    0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Thimerosal, 10 mg/ml Bovine Serum Albumin, Immunoglobulin and Protease free
  • Temperatura di conservazione:
    Cool
  • Concentrazione:
    1,0 mg/ml by UV absorbance at 280 nm
  • Temperatura di spedizione:
    Ice
  • Immunogeno:
    Anti-Ferret IgG IgA and IgM whole molecule was produced by repeated immunization with Ferret IgG IgA and IgM whole molecules in goat.
  • Purificazione:
    purified by immunoaffinity chromatography
  • Taglia:
    1 mg
  • Conf:
    1 mg

Specifications

About this item

Anti-Ferret IgG IgA and IgM whole molecule antibody is suitable for use in immunoelectrophoresis, western-blot, competitive western-blot, ELISA and competitive ELISA assays.

Recommended Dilutions: ELISA: 1:10000 - 1:50000; Western Blot: 1:1000 - 1:5000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgA/IgG/IgM
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Ferret