Order Entry
Switzerland
ContactUsLinkComponent
Anti-VIM Rabbit Polyclonal Antibody
Anti-VIM Rabbit Polyclonal Antibody
Codice catalogo ABCAAB138390-100
Nome del fornitore:  Abcam
Anti-VIM Rabbit Polyclonal Antibody
Codice catalogo ABCAAB138390-100
Nome del fornitore:  Abcam

Specifiche

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    Vimentin
  • Simbolo dell'antigene:
    VIM
  • Clonalità:
    Policlonale
  • Coniugazione:
    Non coniugato
  • Ospite:
    Rabbit
  • Reattività:
    Human
  • Western blot:
    Yes
  • Temperatura di conservazione:
    Cool
  • Temperatura di spedizione:
    Ice
  • Taglia:
    100 µg
  • Conf:
    100 µG

Specifiche

Informazioni su questo articolo

Caution:

All products are FOR RESEARCH USE ONLY.   PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES.  Customer is granted a limited use non-exclusive licence for its own internal research.  Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product.  By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: VIM (vimentin)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human