Inserisci ordine
Switzerland
Orders LinkContactUsLinkComponent
Anti-GluA4 Goat Polyclonal Antibody

Anti-GluA4 Goat Polyclonal Antibody

50 µg, Vial
Codice catalogo ABCAAB115322-50
Marketsource
Nome del fornitore:   Abcam
Anti-GluA4 Goat Polyclonal Antibody
Codice catalogo ABCAAB115322-50
Nome del fornitore:  Abcam
Marketsource
Anti-GluA4 Goat Polyclonal Antibody
Anti-GluA4 Goat Polyclonal Antibody

50 µg, Vial

Price
SFr. 514.00
Log in per disponibilità
1 * 50 µG
Product Details & Documents

Goat polyclonal GluA4 antibody. Suitable for IHC-P, WB and reacts with Human samples. Cited in 4 publications. Immunogen corresponding to Synthetic Peptide within Human GRIA4 aa 250 to 300.

Attenzione: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Primary
Antigen: GluA4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human

Specifiche
  • Seq:
    1202
  • Taglia:
    50 µg
  • Tipo di conf.:
    Vial
  • Western blot:
    Yes
  • Reattività:
    Human
  • Immunogeno:
    Synthetic Peptide within Human GRIA4 aa 250-300. The exact immunogen used to generate this antibody is proprietary information.
  • Purificazione:
    Immunogen Affinity purification
  • Coniugazione:
    Non coniugato
  • Isotipo:
    IgG
  • Peso molecolare:
    100871 Da
  • Cat. No.:
    ABCAAB115322-50
  • Sinomini antigene:
    GRIA4|Glutamate receptor 4|GluR-4|AMPA-selective glutamate receptor 4|GluR-D
  • Forma:
    Liquid
  • Applicazioni testate:
    IHC-P
  • Simbolo dell'antigene:
    GluA4
  • Clonalità:
    Policlonale
  • Ospite:
    Goat
  • Temperatura di spedizione:
    4 °C
  • Tipo anticorpo:
    Primary
  • Temperatura di conservazione:
    Store at 4 °C short term (1 - 2 weeks). Upon delivery aliquot. Store at –20 °C long term.
  • Tipo di anticorpi:
    GLUR4
  • ImmunoChimica:
    Yes
  • Concentrazione:
    0,5 mg/ml