Anti-MTCO2 Mouse Monoclonal Antibody [clone: 4B12A5]
ABCAAB110271-100
: Abcam
- Tipo anticorpo:Primary
- Tipo di anticorpi:Cytochrome C oxidase subunit II
- Clonalità:Monoclonale
- Clone:4B12A5
- Coniugazione:Non coniugato
- Ospite:Mouse
- Isotipo:IgG2a
- Western blot:Yes
- Seq:1286
- Temperatura di conservazione:Cool
- Temperatura di spedizione:Ice
- Taglia:100 µg
- Tipo di conf.:Vial
- Conf:100 µG
Mouse monoclonal [4B12A5] to MTCO2.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: MTCO2
Clonality: Monoclonal
Clone: 4B12A5
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: S. cerevisiae