Anti-TMLHE Rabbit Polyclonal Antibody
ABCAAB97842-50
: Abcam
- Tipo anticorpo:Primary
- Tipo di anticorpi:trimethyllysine hydroxylase, epsilon
- Simbolo dell'antigene:TMLHE
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- Ospite:Rabbit
- Isotipo:IgG
- Reattività:Human
- Western blot:Yes
- Seq:1202
- Temperatura di conservazione:Cool
- Temperatura di spedizione:Ice
- Taglia:50 µl
- Tipo di conf.:Vial
- Conf:50 µl
Rabbit polyclonal to TMLHE.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: TMLHE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human