Anti-UFD1L Rabbit Polyclonal Antibody
Rabbit Polyclonal UFD1L antibody. Suitable for WB, IHC-P, ICC/IF and reacts with Human samples. Cited in 3 publications. Immunogen corresponding to Recombinant Fragment Protein within Human UFD1 aa 1-300.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: UFD1L
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
- Seq:1202
- Taglia:50 µl
- Tipo di conf.:Vial
- Western blot:Yes
- Reattività:Human
- Immunogeno:Recombinant Fragment Protein within Human UFD1 aa 1-300. The exact immunogen used to generate this antibody is proprietary information.
- Purificazione:Immunogen affinity purification
- Tampone di conservazione:Glycerol, tris, Glycine
- Coniugazione:Non coniugato
- Isotipo:IgG
- Peso molecolare:34500 Da
- Cat. No.:ABCAAB96648-50
- Sinomini antigene:Ubiquitin recognition factor in ER-associated degradation protein 1|Ubiquitin fusion degradation protein 1|UB fusion protein 1
- ImmunoFluorescenza:Yes
- Forma:Liquid
- Applicazioni testate:ICC/IF, IHC-P
- Simbolo dell'antigene:UFD1L
- Clonalità:Policlonale
- Ospite:Rabbit
- Temperatura di spedizione:4 °C
- Tipo anticorpo:Primary
- Temperatura di conservazione:Upon delivery aliquot and store at −20 or −80 °C. Avoid repeated freeze/thaw cycles.
- Tipo di anticorpi:UFD1
- ImmunoChimica:Yes
- Concentrazione:1,02 mg/ml
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



