Anti-Flagellin Rabbit Polyclonal Antibody
Codice catalogo ABCAAB93713-100
Nome del fornitore: Abcam
Specifiche
- Tipo anticorpo:Primary
- Tipo di anticorpi:Flagellin
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- Ospite:Rabbit
- ImmunoChimica:Yes
- ImmunoFluorescenza:Yes
- Isotipo:IgG
- Western blot:Yes
- Temperatura di conservazione:Cool
- Temperatura di spedizione:Ice
- Taglia:100 µl
- Tipo di conf.:Vial
- Conf:100 µl
Specifiche
Informazioni su questo articolo
Rabbit polyclonal to Flagellin.
Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: Flagellin
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: E. coli;Salmonella a;Salmonella s;Salmonella