Order Entry
Switzerland
ContactUsLinkComponent
Anti-IgG4 Rabbit Polyclonal Antibody
Anti-IgG4 Rabbit Polyclonal Antibody
Catalog # ABCAAB86251-150
Supplier:  Abcam
undefined
Anti-IgG4 Rabbit Polyclonal Antibody
Catalog # ABCAAB86251-150
Supplier:  Abcam

Specifications

  • Tipo anticorpo:
    Secondary
  • Simbolo dell'antigene:
    IgG4
  • Clonalità:
    Policlonale
  • Coniugazione:
    Non coniugato
  • ELISA:
    Yes
  • Ospite:
    Rabbit
  • Isotipo:
    IgG
  • Western blot:
    Yes
  • Temperatura di conservazione:
    Cool
  • Temperatura di spedizione:
    Ice
  • Taglia:
    150 µg
  • Tipo di conf.:
    Vial
  • Conf:
    150 µG

Specifications

About this item

Rabbit Anti-Human IgG4 H&L.

Caution: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.

Type: Secondary
Antigen: IgG4
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human