- Tipo anticorpo:Primary
- Tipo di anticorpi:Serine/threonine Kinase 3
- Simbolo dell'antigene:STK3
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- Ospite:Rabbit
- ImmunoPrecipitazione:Yes
- Isotipo:IgG
- Reattività:Human,Mouse
- Western blot:Yes
- Seq:1286
- Temperatura di conservazione:Cool
- Temperatura di spedizione:Ice
- Taglia:100 µl
- Tipo di conf.:Vial
- Conf:100 µl
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: STK3 (serine/threonine kinase 3)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse