Anti-CHMP4A Mouse Polyclonal Antibody
Mouse polyclonal CHMP4A antibody. Carrier free. Suitable for WB and reacts with Human samples. Cited in 2 publications. Immunogen corresponding to recombinant Full Length Protein corresponding to Human CHMP4A.
: All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive license for its own internal research. Any control manufacturer supplies with a product is limited to use only to validate the relevant product. By using the product, customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the products or portions thereof.
Type: Primary
Antigen: CHMP4A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
- Seq:1202
- Taglia:50 µg
- Tipo di conf.:Vial
- Western blot:Yes
- Reattività:Human
- Immunogeno:Recombinant Full Length Protein corresponding to Human CHMP4A
- Purificazione:Protein G Affinity purification
- Tampone di conservazione:PBS
- Coniugazione:Non coniugato
- Isotipo:IgG
- Peso molecolare:25098 Da
- Cat. No.:ABCAAB67058-50
- Sinomini antigene:SHAX2|CDA04|HSPC134|Charged multivesicular body protein 4a|Chromatin-modifying protein 4a|SNF7 homolog associated with Alix-2|SNF7-1|Vacuolar protein sorting-associated protein 32-1|hSnf-1|Vps32-1|hVps32-1
- Forma:Liquid
- Simbolo dell'antigene:CHMP4A
- Clonalità:Policlonale
- Ospite:Mouse
- Temperatura di spedizione:4 °C
- Tipo anticorpo:Primary
- Temperatura di conservazione:Upon delivery aliquot and store at −20 or −80 °C. Avoid repeated freeze/thaw cycles.
- Tipo di anticorpi:C14orf123
- Concentrazione:0,5 mg/ml



