Specifiche
- Tipo anticorpo:Primary
- Tipo di anticorpi:v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog
- Simbolo dell'antigene:SRC
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- ELISA:Yes
- Ospite:Rabbit
- ImmunoChimica:Yes
- ImmunoFluorescenza:Yes
- Isotipo:IgG
- Reattività:Pig,Human,Rat,Mouse
- Western blot:Yes
- Temperatura di conservazione:Cool
- Temperatura di spedizione:Ice
- Taglia:100 µg
- Tipo di conf.:Vial
- Conf:100 µG
Specifiche
Informazioni su questo articolo
Rabbit polyclonal to Src.
All products are FOR RESEARCH USE ONLY. PRODUCTS ARE NOT FOR USE IN DIAGNOSTIC OR THERAPEUTIC PROCEDURES. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: SRC (v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Pig, Rat