Anti-CDK11/CDC2L2/CDK11A Rabbit Polyclonal Antibody
Katalognr. ABCAAB19393-100
Anbieter: Abcam
Technische Daten
- Tipo anticorpo:Primary
- Tipo di anticorpi:Cyclin-dependent kinase 11A
- Simbolo dell'antigene:CDK11A
- Clonalità:Policlonale
- Coniugazione:Non coniugato
- Ospite:Rabbit
- ImmunoChimica:Yes
- ImmunoFluorescenza:Yes
- ImmunoPrecipitazione:Yes
- Isotipo:IgG
- Reattività:Human,Mouse
- Western blot:Yes
- Temperatura di conservazione:Cool
- Temperatura di spedizione:Ice
- Taglia:100 µg
- Tipo di conf.:Vial
- Conf:100 µG
Technische Daten
Über diesen Artikel
Rabbit polyclonal to CDK11/CDC2L2/CDK11A.
Caution: All products are for research use only. Products are not for use in diagnostic or therapeutic procedures. Customer is granted a limited use non-exclusive licence for its own internal research. Any control manufacturer supplies with a Product is limited to use only to validate the relevant Product. By using the Product, Customers agree not to attempt to reverse engineer or otherwise perform any compositional, structural, functional or other analyses directed to learning the methodology, formulae, sequences, processes, make-up or production of any of the Products or portions thereof.
Type: Primary
Antigen: CDK11/CDC2L2/CDK11A
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse