Anti-IgM Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BIRBORB21624-1
Supplier: Biorbyt
Specifications
- Tipo anticorpo:Secondary
- Simbolo dell'antigene:IgM
- Clonalità:Policlonale
- Coniugazione:FITC (Fluoresceina Isotiocionato)
- Ospite:Rabbit
- Formato:immunoconjugate
- Forma:lyophilized
- Temperatura di conservazione:Cool
- Concentrazione:1 mg/ml, Fluorochrome/IgG protein molar ratio (F/P) is approximately 1,3, no foreign proteins added,
- Temperatura di spedizione:Ice
- Immunogeno:Purified homogenous IgM isolated from pooled mouse serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Taglia:1 ml
- Conf:1 mL
Specifications
About this item
Type: Secondary
Antigen: IgM (Mouse) (mouse IgM)
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: none
Reactivity: