Anti-IgA Goat Polyclonal Antibody (TRITC (Tetramethylrhodamine Isothiocyanate))
Catalog # BIRBORB21600-1
Supplier: Biorbyt
Specifications
- Tipo anticorpo:Secondary
- Simbolo dell'antigene:IgA
- Clonalità:Policlonale
- Coniugazione:TRITC (Tetrametillrodamina Isotiocianato)
- Ospite:Goat
- Formato:immunoconjugate
- Forma:lyophilized
- Temperatura di conservazione:Cool
- Concentrazione:1 mg/ml, Fluorochrome/IgG protein molar ratio (F/P) is approximately 1,3, no foreign proteins added,
- Temperatura di spedizione:Ice
- Immunogeno:Purified polyclonal IgA isolated from pooled Rhesus monkey serum. Freunds complete adjuvant is used in the first step of the immunization procedure.
- Taglia:1 ml
- Conf:1 mL
Specifications
About this item
Type: Secondary
Antigen: Monkey IgA
Clonality: polyclonal
Clone:
Conjugation: TRITC (Rhodamine)
Epitope:
Host: Goat
Isotype: none
Reactivity: