Order Entry
Switzerland
ContactUsLinkComponent
 
Anti-PKA gamma Rabbit Polyclonal Antibody (FITC (Fluorescein))
  BIRBORB9578-100
 :  Biorbyt
Anti-PKA gamma Rabbit Polyclonal Antibody (FITC (Fluorescein))
  BIRBORB9578-100
 :  Biorbyt

 

  • Tipo anticorpo:
    Primary
  • Simbolo dell'antigene:
    PKA gamma
  • Clonalità:
    Policlonale
  • Coniugazione:
    FITC (Fluoresceina Isotiocionato)
  • Ospite:
    Rabbit
  • ImmunoFluorescenza:
    Yes
  • Isotipo:
    IgG
  • Reattività:
    Human,
    Rat,
    Mouse
  • Forma:
    liquid
  • Tampone di conservazione:
    10 mM PBS, pH 7,4 with 10 mg/ml BSA, 0,03% Proclin 300 and 25% glycerol
  • Peso molecolare:
    40 kDa
  • Temperatura di conservazione:
    Cool
  • Concentrazione:
    0,5 mg/ml
  • Temperatura di spedizione:
    Ice
  • Immunogeno:
    KLH conjugated synthetic peptide derived from human PKA gamma.
  • Purificazione:
    purified by affinity chromatography
  • Taglia:
    100 µg
  • Conf:
    100 µG

 

 

Immunofluorescence: 1:50-200 (based on 0.5 mg/ml)

Type: Primary
Antigen: PKA gamma
Clonality: polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat