Anti-FCGR2A Rabbit Polyclonal Antibody [clone: 439CT10.4.4]
Catalog # ABGEAP8918B
Supplier: Abgent
Specifications
- Tipo anticorpo:Primary
- Tipo di anticorpi:Nuclear receptor coactivator 4
- Simbolo dell'antigene:NCOA4
- Clonalità:Policlonale
- Clone:439CT10.4.4
- Coniugazione:Non coniugato
- ELISA:Yes
- Citometria a flusso:Yes
- Ospite:Rabbit
- Isotipo:IgM
- Reattività:Human
- Western blot:Yes
- Epitopo:C-terminal
- Forma:Ascites
- ID gene:8031
- Sinomini antigene:CD32|FcGR|CD32A|IGFR2|FCG2|FCGR2A1|CDw32|FCGR2|FCGR2A
- Tampone di conservazione:PBS with 0,09% (W/V) sodium azide, pH 7,4
- Sequenza:EEVCRANEPCTSFAECVCDENCEKEALYKWLLKKEGKDKNGMPVEPKPEPEKHKDSLNMWLCPRKEVIEQTKAPKAMTPSRIADSFQVIKNSPLSEWLIRPPYKEGSPKEVPGTEDRAGKQKFKSPMNTSWCSFNTADWVLPGKKMGNLSQLSSGEDKWLLRKKAQEVLLNSPLQEEHNFPPDHYGLPAVCDLFACMQLKVDKEKWLYRTPLQ
- Temperatura di conservazione:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Temperatura di spedizione:4 °C
- Immunogeno:Purified His-tagged NCOA4 protein(Fragment) was used to produced this monoclonal antibody.
- Conf:400 µl
Specifications
About this item
Type: Primary
Antigen: FCGR2A (Fc fragment of IgG, low affinity IIa, receptor (CD32))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human