Order Entry
Switzerland
ContactUsLinkComponent
Anti-LPIN2 Rabbit Polyclonal Antibody
Anti-LPIN2 Rabbit Polyclonal Antibody
Catalog # ABGEAP8583C
Supplier:  Abgent
undefined
Anti-LPIN2 Rabbit Polyclonal Antibody
Catalog # ABGEAP8583C
Supplier:  Abgent

Specifications

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    Lipin 2
  • Simbolo dell'antigene:
    LPIN2
  • Clonalità:
    Policlonale
  • Coniugazione:
    Non coniugato
  • ELISA:
    Yes
  • Citometria a flusso:
    Yes
  • Ospite:
    Rabbit
  • ImmunoChimica:
    Yes
  • Reattività:
    Human
  • Western blot:
    Yes
  • Epitopo:
    Center
  • ID gene:
    970
  • Sinomini antigene:
    EC 3.1.3.4|KIAA0249
  • Tampone di conservazione:
    PBS with 0,09% (W/V) sodium azide, pH 7,4
  • Sequenza:
    HGPELDKGQLRIHRDC
  • Temperatura di conservazione:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Concentrazione:
    0,5 mg/ml
  • Temperatura di spedizione:
    4 °C
  • Immunogeno:
    This CD70 antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 72-101 amino acids from the Central region of human CD70.
  • Purificazione:
    Protein A & Peptide affinity purification
  • Conf:
    400 µl

Specifications

About this item

Type: Primary
Antigen: LPIN2 (lipin 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: middle
Host: Rabbit
Isotype: none
Reactivity: Human