Order Entry
Switzerland
ContactUsLinkComponent
Anti-UFC1 Rabbit Polyclonal Antibody
Anti-UFC1 Rabbit Polyclonal Antibody
  ABGEAP2187B
 :  Abgent
undefined
Anti-UFC1 Rabbit Polyclonal Antibody
  ABGEAP2187B
 :  Abgent

 

  • Tipo anticorpo:
    Primary
  • Tipo di anticorpi:
    Ubiquitin-fold modifier conjugating enzyme 1
  • Simbolo dell'antigene:
    UFC1
  • Clonalità:
    Policlonale
  • Coniugazione:
    Non coniugato
  • ELISA:
    Yes
  • Ospite:
    Rabbit
  • Reattività:
    Human,
    Mouse
  • Western blot:
    Yes
  • Epitopo:
    C-Terminal
  • Assorbimento incrociato:
    No
  • Sinomini antigene:
    Ubiquitin-fold modier-conjugating enzyme 1|UFC1_HUMAN|CGI-126|Ubiquitin fold modier conjugating enzyme 1|UFC1|HSPC155|Ufm1 conjugating enzyme 1|Ufm1-conjugating enzyme 1.
  • Temperatura di conservazione:
    Cool
  • Temperatura di spedizione:
    Ice
  • Conf:
    400 µl

 

 

Type: Primary
Antigen: UFC1 (ubiquitin-fold modifier conjugating enzyme 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope: C-terminal
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse